Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z742677

Artis Tweezer

Style 7-SA

Synonym(s):

Artis Tweezer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
27112105
NACRES:
MA.01

feature

tip style (Curved, super fine)

manufacturer/tradename

Aven 18072ARS

technique(s)

PCR: suitable

L

115 mm (4.5 in.)

Looking for similar products? Visit Product Comparison Guide

General description

Aven′s innovative Artis Tweezers have set a new precedent for precision. Our cutting edge design features a angled bend at the end of the tip, which provides a softer overall grip with a large surface area that reduces hand fatigue, enhancing productivity.

The durable tweezers are protected by an attractive ESD safe coating, and the highly polished stainless steel tips with rounded edges allow operators to handle very fine wire, miniature parts, electrical components, and other delicate materials in a confined space without risk of damage.

Application

Curved, super fine points for assembly work

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service