Key Documents
Z680222
Titanium tweezers
Style 3
About This Item
UNSPSC Code:
27112105
NACRES:
MA.01
manufacturer/tradename
Aven
total L
4.75 in. (121 mm)
General description
Titanium tweezers do not shed particles like stainless steel tweezers. Titanium is light weight, high strength, non-magnetic material.
  • Lightweight (high strength to weight ratio)
  • Zero magnetic retentively
  • Temperature resistant to 800 °F
  • Chemical and corrosion resistant
  • Non-solder adhesive
  • Bio-compatible (maintain cell integrity, no inflammatory response)
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)