Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22435-U

Sigma-Aldrich

Stainless Steel Tweezers

L 6 in., pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
27112105
NACRES:
NB.35

form

solid

packaging

pkg of 1 ea

L

6 in.

General description

Stainless Steel Tweezers - Perfect for picking up hot injector liners and detector parts or for preventing contamination with finger oils. Also useful for holding ferrules while inserting capillary tubing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service