Skip to Content
MilliporeSigma
All Photos(1)

Documents

D161101

Sigma-Aldrich

N,N′-Dimethyl-1,6-hexanediamine

98%

Synonym(s):

1,6-Bis(methylamino)hexane, N,N′-Dimethyl-1,6-diaminohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH(CH2)6NHCH3
CAS Number:
Molecular Weight:
144.26
Beilstein:
1735385
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

96 °C/14 mmHg (lit.)

mp

15-18 °C (lit.)

density

0.807 g/mL at 25 °C (lit.)

SMILES string

CNCCCCCCNC

InChI

1S/C8H20N2/c1-9-7-5-3-4-6-8-10-2/h9-10H,3-8H2,1-2H3

InChI key

MDKQJOKKKZNQDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N′-Dimethyl-1,6-hexanediamine (DMHDA) can be used as an organic building block to synthesize:
  • Hydroxyurethane by reacting with propylene carbonate.
  • N,N′-dimethyl, N,N′-di(methacryloxy ethyl)-1,6-hexanediamine (NDMH), a monomer applicable as both a co-initiator and a reactive diluent for dental resins.
  • Degradable poly(amino alcohol ester) polymer by nucleophilic addition with diglycidyl adipate for the potential use in gene transfection vectors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactivity of secondary amines for the synthesis of non-isocyanate polyurethanes
Camara F, et al.
European Polymer Journal, 55, 17-26 (2014)
Synthesis and photopolymerization of N, N?-dimethyl,-N, N?-di (methacryloxy ethyl)-1, 6-hexanediamine as a polymerizable amine coinitiator for dental restorations
Nie J and Bowman CN
Biomaterials, 23(4), 1221-1226 (2002)
Degradable poly (amino alcohol esters) as potential DNA vectors with low cytotoxicity
J Sangyong, et al.
Biomacromolecules, 4(6), 1759-1762 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service