Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D13208

Sigma-Aldrich

1,4-Diaminobutane

99%

Synonym(s):

1,4-Butanediamine, Putrescine, Tetramethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4NH2
CAS Number:
Molecular Weight:
88.15
Beilstein:
605282
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

expl. lim.

9.08 %

refractive index

n20/D 1.457 (lit.)

bp

158-160 °C (lit.)

mp

25-28 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

SMILES string

NCCCCN

InChI

1S/C4H12N2/c5-3-1-2-4-6/h1-6H2

InChI key

KIDHWZJUCRJVML-UHFFFAOYSA-N

Gene Information

rat ... Odc1(24609)

Looking for similar products? Visit Product Comparison Guide

Application

GABA precursor in many biological systems[1] and synthon for amido-ureas.[2]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 3179-3179 (1992)
M C de Mello et al.
Neurochemistry international, 22(3), 249-253 (1993-03-01)
Cultured retina cells from chick embryos took up [3H]putrescine and approx 10.8% of the incorporated amine was converted into [3H]GABA. The putrescine-derived GABA accumulated in a pool that was released in the medium at a rate corresponding to 3.66% of
Juhani Jänne et al.
European journal of biochemistry, 271(5), 877-894 (2004-03-11)
The polyamines putrescine, spermidine and spermine are organic cations shown to participate in a bewildering number of cellular reactions, yet their exact functions in intermediary metabolism and specific interactions with cellular components remain largely elusive. Pharmacological interventions have demonstrated convincingly
Chi Yang et al.
PloS one, 7(10), e47314-e47314 (2012-10-17)
The phosphate starvation response in bacteria has been studied extensively for the past few decades and the phosphate-limiting signal is known to be mediated via the PhoBR two-component system. However, the global DNA binding profile of the response regulator PhoB
A Raina et al.
Medical biology, 53(3), 121-147 (1975-06-01)
The biochemistry and biological function of the naturally occurring polyamines, putrescine, spermidine, and spermine, have been reviewed with special reference to animal organisms. These compounds are universally distributed in all living material. Their biosynthesis from ornithine and methionine is accurately

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service