Skip to Content
MilliporeSigma

8.04323

1,6-Diaminohexane

for synthesis

Synonym(s):

1,6-Diaminohexane, 1,6-Hexanediamine, Hexamethylenediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H2N(CH2)6NH2
CAS Number:
Molecular Weight:
116.20
EC Index Number:
204-679-6
UNSPSC Code:
12352116
MDL number:
Assay:
≥99% (GC)
Grade:
synthesis grade
Bp:
199-204 °C/1013 hPa
Vapor pressure:
0.25 hPa ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

vapor pressure

0.25 hPa ( 20 °C)

assay

≥99% (GC)

form

solid

autoignition temp.

305 °C

potency

850 mg/kg LD50, oral (Rat), 1110 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-7.6 % (v/v)

dilution

(for synthesis)

pH

12.4 (25 °C, 100 g/L in H2O)

bp

199-204 °C/1013 hPa

mp

39-42 °C

transition temp

flash point 85 °C

solubility

490 g/L

density

0.83 g/cm3 at 60 °C

storage temp.

2-30°C

InChI

1S/C6H16N2/c7-5-3-1-2-4-6-8/h1-8H2

InChI key

NAQMVNRVTILPCV-UHFFFAOYSA-N

Application

  • Antibacterial Activity of Schiff Bases Derived from 1,6-Diaminohexane: Discusses the synthesis and antibacterial properties of Schiff bases derived from 1,6-diaminohexane, presenting a QSAR study with quantum mechanics (NJB Kangah et al., 2017).


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

wgk

WGK 1

flash_point_f

176.0 °F - closed cup

flash_point_c

80 °C - closed cup

Storage Class

8A - Combustible corrosive hazardous materials



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library