Skip to Content
MilliporeSigma
N,N′-Dimethyl-1,6-hexanediamine 98%

N,N′-Dimethyl-1,6-hexanediamine

N,N′-Dimethyl-1,6-hexanediamine 98%
Synonym(s):
1,6-Bis(methylamino)hexane, N,N′-Dimethyl-1,6-diaminohexane
Linear Formula:
CH3NH(CH2)6NHCH3
CAS Number:
Molecular Weight:
144.26
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing