Skip to Content
MilliporeSigma
All Photos(3)

Documents

105139

Sigma-Aldrich

N,N,N′,N′-Tetramethyl-1,6-hexanediamine

99%

Synonym(s):

1,1,8,8-Tetramethyl-1,8-diazaoctane, 1,6-Bis(dimethylamino)hexane, 2,9-Dimethyl-2,9-diazadecane, N ,N ,N ′,N ′-Tetramethylhexylenediamine, Hexamethylenebis(dimethylamine), N 1 ,N 1 ,N 6 ,N 6 -Tetramethyl-1,6-hexanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2N(CH2)6N(CH3)2
CAS Number:
Molecular Weight:
172.31
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

refractive index

n20/D 1.4359 (lit.)

bp

209-210 °C (lit.)

density

0.806 g/mL at 25 °C (lit.)

SMILES string

CN(C)CCCCCCN(C)C

InChI

1S/C10H24N2/c1-11(2)9-7-5-6-8-10-12(3)4/h5-10H2,1-4H3

InChI key

TXXWBTOATXBWDR-UHFFFAOYSA-N

Application

N,N,N′,N′-Tetramethyl-1,6-hexanediamine (TMHDA)can be used as an aminating agent to prepare anion exchange membranes for solidalkaline fuel cells. It provides cationic groups for better anionic conductivityand acts as a cross-linking agent to enhance dimensional stability. Theaddition of TMHDA combines functionalizing and crosslinking processes into asingle step.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingqian Meng et al.
ChemCatChem, 9(20), 3942-3954 (2017-12-05)
The influence of framework substituents (Al
Volkan Şahin et al.
Nanoscale, 12(30), 16292-16304 (2020-07-29)
Understanding the chemical mechanism of crystal nucleation at the molecular level is crucial for the design of architectural structures of valuable materials in the future. In this study, it has been revealed that amorphous silicate precursors, which play a role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service