Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808555

Sigma-Aldrich

N-Butyl phosphate, mixture of mono-N-Butyl and di-N-Butyl phosphate

95%

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

transition temp

flash point 230 °F

density

1.123 g/mL at 25 °C

functional group

phosphate

SMILES string

[P](=O)(OCCCC)(O)O

InChI

1S/C4H11O4P/c1-2-3-4-8-9(5,6)7/h2-4H2,1H3,(H2,5,6,7)

InChI key

BNMJSBUIDQYHIN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service