Skip to Content
MilliporeSigma
All Photos(3)

Documents

737933

Sigma-Aldrich

Butylphosphonic acid

Synonym(s):

n-Butylphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11O3P
CAS Number:
Molecular Weight:
138.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

98-102 °C

SMILES string

CCCCP(O)(O)=O

InChI

1S/C4H11O3P/c1-2-3-4-8(5,6)7/h2-4H2,1H3,(H2,5,6,7)

InChI key

UOKRBSXOBUKDGE-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Dumora et al.
The Biochemical journal, 280 ( Pt 2), 557-559 (1991-12-01)
The effect of n-butylphosphonic acid on the activity of phosphonoacetaldehyde hydrolase from Pseudomonas aeruginosa was investigated: at low concentrations this compound appeared as an activator of the enzyme activity, whereas at higher concentrations it exhibited inhibitory properties. The experimental results
Chung-Wei Yang et al.
Scientific reports, 7(1), 16910-16910 (2017-12-06)
The hydrothermal treatment followed by a self-assembled monolayer (SAM) of 1-butylphosphonic acid through the tethering by aggregation and growth (T-BAG) method was employed to produce protective surface coatings on the Mg-6Al-1Zn alloy (AZ61) for reducing the degradation rate in physiological
N Amend et al.
Archives of toxicology, 94(6), 2239-2247 (2020-04-19)
Suicidal ingestion of organophosphorus (OP) or carbamate (CM) compounds challenges health care systems worldwide, particularly in Southeast Asia. The diagnosis and treatment of OP or CM poisoning is traditionally based on the clinical appearance of the typical cholinergic toxidrome, e.g.

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service