Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.20251

Sigma-Aldrich

Butyl phosphate

(mixture of mono and di esters) for synthesis

Synonym(s):

Dibutyl phosphate, Phosphoric acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

bp

>145 °C/1013 hPa

transition temp

flash point >108.5 °C

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

227.3 °F

Flash Point(C)

> 108.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service