Skip to Content
MilliporeSigma

1.00002

Tri-n-butyl phosphate

EMPROVE® EXPERT, Ph. Eur.

Synonym(s):

Tributyl phosphate, TBP, TBPA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
EC Index Number:
204-800-2
UNSPSC Code:
12352303
Beilstein/REAXYS Number:
1710584
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

Ph. Eur.

Quality Segment

vapor density

9.2 (vs air)

vapor pressure

27 mmHg ( 178 °C), 7.3 mmHg ( 150 °C)

product line

EMPROVE® EXPERT

form

liquid

autoignition temp.

770 °F

refractive index

n20/D 1.424 (lit.)

bp

180-183 °C/22 mmHg (lit.)

mp

−79 °C (lit.)

transition temp

flash point 146 °C

solubility

6 g/L

density

0.979 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical

storage temp.

15-25°C

SMILES string

CCCCOP(=O)(OCCCC)OCCCC

InChI

1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

STCOOQWBFONSKY-UHFFFAOYSA-N

General description

Chemicals play an important role in the stabilization of a biologic drug during its manufacturing and formulation process – for instance, by preventing aggregation. We offer a wide range of high-quality stabilizers, buffers and salts to successfully purify and formulate your biomolecules. Specifically developed for high-risk applications, they are low in bioburden and endotoxins.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Tri-n-butyl phosphate is used widely as a solvent along with either TritonTM X100 or Tween® in solvent detergent treatment for viral inactivation procedures especially for plasma derived therapeutics.
Due to its low microbial and endotoxin limits, Tri-n-butyl phosphate Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC


Still not finding the right product?

Explore all of our products under Tri-n-butyl phosphate


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

293.0 °F - closed cup

flash_point_c

145 °C - closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
100002902604027269080471
100002050004027269080457
100002250004027269080464