Skip to Content
MilliporeSigma
All Photos(3)

Documents

198781

Sigma-Aldrich

12-Hydroxydodecanoic acid

97%

Synonym(s):

12-Hydroxylauric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)11COOH
CAS Number:
Molecular Weight:
216.32
Beilstein:
1238370
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

85-88 °C (lit.)

SMILES string

OCCCCCCCCCCCC(O)=O

InChI

1S/C12H24O3/c13-11-9-7-5-3-1-2-4-6-8-10-12(14)15/h13H,1-11H2,(H,14,15)

InChI key

ZDHCZVWCTKTBRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

12-Hydroxydodecanoic acid was used in the synthesis of high molecular weight poly[(12-hydroxydodecanoate)-co-(12-hydroxystearate)] [poly(12HD-co-12HS)] samples with variable monomer ratios using methyl 12-hydroxystearate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Bailo et al.
Analytical and bioanalytical chemistry, 394(7), 1797-1801 (2009-06-16)
Surface-enhanced Raman scattering was used as a spectroscopic tool to investigate the changes brought upon cytochrome P450BSss after fatty acid binding. Differences in the spectra of substrate-free and substrate-bound enzyme were observed indicating the potential for this method to be
Alexander N Grechkin et al.
FEBS letters, 549(1-3), 31-34 (2003-08-14)
Guava (Psidium guajava) hydroperoxide lyase (HPL) preparations were incubated with [1-(14)C](9Z,11E,13S,15Z)-13-hydroperoxy-9,11,15-octadecatrienoic acid for 1 min at 0 degrees C, followed by rapid extraction/trimethylsilylation. Analysis of the trimethylsilylated products by gas chromatography-mass spectrometry and radio-high-performance liquid chromatography revealed a single predominant
H A Dirven et al.
Journal of chromatography, 564(1), 266-271 (1991-03-08)
The formation of omega-hydroxylauric acid from lauric acid is an indicator of the activity of cytochrome P-450 IV family proteins. The two main metabolites of lauric acid, (omega-1)-and omega-hydroxylauric acid, have been completely separated by reversed-phase high-performance liquid chromatography. Measurement
P Jezek et al.
FEBS letters, 408(2), 166-170 (1997-05-19)
Fatty acid (FA) uniport via mitochondrial uncoupling protein (UcP) was detected fluorometrically with PBFI, potassium-binding benzofuran phthalate and SPQ, 6-methoxy-N-(3-sulfopropyl)-quinolinium, indicating K+ and H+, respectively. The FA structural patterns required for FA flip-flop, UcP-mediated FA uniport, activation of UcP-mediated H+
D D Giera et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 16(2), 348-355 (1991-02-01)
Assessment of hepatic omega-oxidation of fatty acids by cytochrome P450IV enzymes in toxicology studies can be a means of evaluating test compound effects on peroxisomal proliferation. Routine assay of omega-oxidation, however, requires a simpler method of enzymatic analysis than currently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service