Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

795658

Sigma-Aldrich

Garg 3,4-pyridyne precursor

97%

Synonym(s):

4-(Trimethylsilyl)pyridin-3-yl trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12F3NO3SSi
CAS Number:
Molecular Weight:
299.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n/D 1.455

density

1.259 at 25 °C

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C1=C(OS(=O)(C(F)(F)F)=O)C=NC=C1

InChI

1S/C9H12F3NO3SSi/c1-18(2,3)8-4-5-13-6-7(8)16-17(14,15)9(10,11)12/h4-6H,1-3H3

InChI key

HJIBLRJECSPWKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pyridyne precursor can be activated under mild fluoride conditions; which are highly reactive with dienes and other dipolariphiles (e.g. azides) to provide cycloaddition products containing an indole motif.

Packaging

Each AldrichCPR product will be packaged in one consolidated unit unless you specify multiple units at the time of order.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

Related Content

The Garg group develops methods for the synthesis of natural products and pharmaceuticals. One key method pertains to heterocyclic arynes, such as indolynes and pyridynes, which are generated in situ from silyltriflate precursors under mild fluoride based reaction conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service