Skip to Content
MilliporeSigma
All Photos(1)

Documents

419982

Sigma-Aldrich

Triethyl(trifluoromethyl)silane

98%

Synonym(s):

(Trifluoromethyl)triethylsilane, Triethylsilyl trifluoromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3SiCF3
CAS Number:
Molecular Weight:
184.27
Beilstein:
4242161
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.382 (lit.)

bp

56-57 °C/60 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

SMILES string

CC[Si](CC)(CC)C(F)(F)F

InChI

1S/C7H15F3Si/c1-4-11(5-2,6-3)7(8,9)10/h4-6H2,1-3H3

InChI key

ZHSKFONQCREGOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Trifluoromethylation of aryl iodides
  • Trialkylsilylation reactions

  • Used for deposition of perfluoro-methyl silica films

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

related product

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

32.0 °F - closed cup

Flash Point(C)

0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Jin Cho et al.
Science (New York, N.Y.), 328(5986), 1679-1681 (2010-06-26)
The trifluoromethyl group can dramatically influence the properties of organic molecules, thereby increasing their applicability as pharmaceuticals, agrochemicals, or building blocks for organic materials. Despite the importance of this substituent, no general method exists for its installment onto functionalized aromatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service