Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

903183

Sigma-Aldrich

Benzyl 4-oxo-3-(triethylsilyl)piperidine-1-carboxylate

≥95%

Synonym(s):

Garg silyl triflate precursor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H29NO3Si
CAS Number:
Molecular Weight:
347.52
UNSPSC Code:
12352005

Assay

≥95%

form

liquid

refractive index

n/D 1.528

density

1.082 g/mL

functional group

ketone
phenyl

storage temp.

−20°C

Application

Benzyl 4-oxo-3-(triethylsilyl)piperidine-1-carboxylate is a precursor to a Garg silyl triflate which upon exposure to fluoride generates a reactive azacyclic allene for cycloaddition reactions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

257.9 °F

Flash Point(C)

125.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joyann S Barber et al.
Nature chemistry, 10(9), 953-960 (2018-08-01)
For over a century, the structures and reactivities of strained organic compounds have captivated the chemical community. Whereas triple-bond-containing strained intermediates have been well studied, cyclic allenes have received far less attention. Additionally, studies of cyclic allenes that bear heteroatoms

Related Content

The Garg group develops methods for the synthesis of natural products and pharmaceuticals. One key method pertains to heterocyclic arynes, such as indolynes and pyridynes, which are generated in situ from silyltriflate precursors under mild fluoride based reaction conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service