Skip to Content
MilliporeSigma
Garg 3,4-pyridyne precursor 97%

Garg 3,4-pyridyne precursor

Garg 3,4-pyridyne precursor 97%
Synonym(s):
4-(Trimethylsilyl)pyridin-3-yl trifluoromethanesulfonate
Empirical Formula (Hill Notation):
C9H12F3NO3SSi
CAS Number:
Molecular Weight:
299.34

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing