Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

795674

Sigma-Aldrich

3-(Trimethylsilyl)pyridin-2-yl trifluoromethanesulfonate

95%

Synonym(s):

Garg 2,3-pyridyne precursor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12F3NO3SSi
CAS Number:
Molecular Weight:
299.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n/D 1.454

density

1.266 at 25 °C

storage temp.

2-8°C

InChI

1S/C9H12F3NO3SSi/c1-18(2,3)7-5-4-6-13-8(7)16-17(14,15)9(10,11)12/h4-6H,1-3H3

InChI key

SXIBFUWSBRRXRJ-UHFFFAOYSA-N

Application

Pyridyne precursor can be activated under mild fluoride conditions; which are highly reactive with dienes and other dipolariphiles (e.g. azides) to provide cycloaddition products containing an indole motif.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

New and efficient methods for the synthesis of functionalized heterocycles are highly sought after.

Related Content

The Garg group develops methods for the synthesis of natural products and pharmaceuticals. One key method pertains to heterocyclic arynes, such as indolynes and pyridynes, which are generated in situ from silyltriflate precursors under mild fluoride based reaction conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service