Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

567671

Sigma-Aldrich

Dichlorobis(trimethylphosphine)nickel(II)

97%

Synonym(s):

Bis(trimethylphosphine)nickel dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[((CH3)3P)]2NICl2
CAS Number:
Molecular Weight:
281.75
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: nickel
reagent type: catalyst

mp

194-199 °C (lit.)

λmax

244 nm

SMILES string

Cl[Ni]Cl.CP(C)C.CP(C)C

InChI

1S/2C3H9P.2ClH.Ni/c2*1-4(2)3;;;/h2*1-3H3;2*1H;/q;;;;+2/p-2

InChI key

KYHNNWWCXIOTKC-UHFFFAOYSA-L

Application

Catalyst for:
  • Wenkert arylation of thiophene with aryl Grignard reagents
  • Regioselective [2+2+2] cycloaddition of carboryne with alkynes to give benzocarborane compounds
  • Kumada-Corriu cross coupling of Grignard reagents
  • Borylation of aryl chlorides
  • Reductive aldol cyclization-lactonization
  • Arylcyanation of alkynes
  • Alkynylation of benzonitriles via C-C bond activation
Used as a catalyst for the cross-coupling of aryl Grignards with benzonitriles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service