Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

406341

Sigma-Aldrich

Nickel(II) bromide ethylene glycol dimethyl ether complex

97%

Synonym(s):

(1,2-Dimethoxyethane)nickel dibromide, Dibromo(1,2-dimethoxyethane)nickel, Dibromo(1,2-dimethoxyethane)nickel(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiBr2 · CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
308.62
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
97%
form:
powder

Quality Level

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: nickel

SMILES string

Br[Ni]Br.COCCOC

InChI

1S/C4H10O2.2BrH.Ni/c1-5-3-4-6-2;;;/h3-4H2,1-2H3;2*1H;/q;;;+2/p-2

InChI key

VHSVJTYBTJCDFL-UHFFFAOYSA-L

General description

Nickel(II) bromide ethylene glycol dimethyl ether complex, also known as Ni(II) bromide ethylene glycol dimethyl ether complex. It is an organometallic coordination compound used as a precursor for the deposition of nickel-containing thin films. It is also found applications in organic synthesis as a Lewis acid catalyst.

Application

Nickel(II) bromide ethylene glycol dimethyl ether complex can be used as a precursor for synthesizing various nickel(II) complexes. These complexes used as a catalysts for:      
  • The selective transformation of ethylene into higher-order products.     
  • The vinyl polymerization of norbornene.      
  • The preparation of poly(3-hexylthiophene) (P3HT) Kumada catalyst transfer polymerization. It facilitates good control over the molecular weight of the conjugating polymer.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 Inhalation - Flam. Sol. 1 - Muta. 2 - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract,Skin

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

26.6 °F - Non-equilibrium method

Flash Point(C)

-3 °C - Non-equilibrium method

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid flow-based synthesis of poly (3-hexylthiophene) using 2-methyltetrahydrofuran as a bio-derived reaction solvent
Bannock JH, et al.
European Polymer Journal, 80, 240-246 (2016)

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service