Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

261971

Sigma-Aldrich

Tricyclohexylphosphine

Synonym(s):

PCy3, P(Cy)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)3P
CAS Number:
Molecular Weight:
280.43
EC Number:
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Hiyama Coupling

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

81-83 °C (lit.)

functional group

phosphine

SMILES string

C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

InChI

1S/C18H33P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h16-18H,1-15H2

InChI key

WLPUWLXVBWGYMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ligand used in the Pd-catalyzed coupling of malononitrile with aryl halides.[1] Bulky phosphine ligand used with a Pd(0)-triolefinic macrocycle catalyst for Suzuki coupling of aryl bromides and chlorides.[2]
This ligand is applied with Ni as a key intermediate in the formation of cylcopentane compounds.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synlett, 3167-3167 (2006)
Sensuke Ogoshi et al.
Journal of the American Chemical Society, 128(16), 5350-5351 (2006-04-20)
Cyclopropyl phenyl ketone underwent oxidative addition to Ni(PCy3) generated from Ni(cod)2 and PCy3 to give a nickeladihydropyran, which is a key intermediate for the Ni(0)-catalyzed homo- or heterocycloaddition to give cyclopentane compounds having two carbonyl substituents at the 1,3-position.
Synlett, 3001-3001 (2006)
Adrian Tlahuext-Aca et al.
Dalton transactions (Cambridge, England : 2003), 43(42), 15997-16005 (2014-09-19)
Ni(0)-catalyzed dehydrogenation of benzylic-type imines was performed to yield asymmetrical tetra-substituted imidazoles and 2-imidazolines. This was achieved with a single operational step while maintaining good selectivity and atom economy. The catalytic system shows low to moderate tolerance for fluoro-, trifluoromethyl-
Sebastijan Kovačič et al.
Macromolecular rapid communications, 36(17), 1605-1611 (2015-07-15)
Metal-organic frameworks (MOFs) nanoparticles in combination with a nonionic surfactant (Pluronic L-121) are used to stabilize dicyclopentadiene (DCPD)-in-water high internal phase emulsions (HIPEs). The resulting HIPEs containing the MIL-100(Fe) nanoparticles (MIL: Materials of Institut Lavoisier) at the interface between the

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service