Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

708526

Sigma-Aldrich

Bis(tricyclohexylphosphine)nickel(II) dichloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H66Cl2NiP2
CAS Number:
Molecular Weight:
690.46
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

reaction suitability

core: nickel
reagent type: catalyst

mp

284 °C

SMILES string

Cl[Ni]Cl.C1CCC(CC1)P(C2CCCCC2)C3CCCCC3.C4CCC(CC4)P(C5CCCCC5)C6CCCCC6

InChI

1S/2C18H33P.2ClH.Ni/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*16-18H,1-15H2;2*1H;/q;;;;+2/p-2

InChI key

YOCBOYPGZVFUCQ-UHFFFAOYSA-L

Application

Catalyst for:
  • Dehydrobrominative polycondensation
  • Arylation reactions
  • Cross-coupling
  • Suzuki-miyaura cross-coupling reactions
  • Kumada coupling of diaryl sulfates with Grignard reagents
  • Olefin dimerization

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service