Skip to Content
MilliporeSigma
All Photos(2)

Documents

328219

Sigma-Aldrich

[1,2-Bis(diphenylphosphino)ethane]dichloronickel(II)

Synonym(s):

1,2-Ethylenebis(diphenylphosphine)nickel dichloride, Nickel 1,2-Bis(diphenylphosphino)ethane dichloride, [1,2-Bis(diphenylphosphino)ethane]nickel(II) chloride, dppeNiCl2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)2PCH2CH2P(C6H5)2]NiCl2
CAS Number:
Molecular Weight:
528.02
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: nickel
reagent type: catalyst

mp

263-265 °C (lit.)

SMILES string

Cl[Ni]Cl.C(CP(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C26H24P2.2ClH.Ni/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)21-22-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26;;;/h1-20H,21-22H2;2*1H;/q;;;+2/p-2

InChI key

XXECWTBMGGXMKP-UHFFFAOYSA-L

Application

Reactant involved in:
  • Kumada catalyst-transfer polycondensation
  • Oxidation of carboranyl phosphine ligands
  • Synthesis of nickel-iron dithiolato hydrides
  • Ullmann reactions - homocoupling reactions

Co-catalyst for hydroformylation of alcohols for the synthesis of quaternary carbon centers

Catalyst for Wacker-type intramolecular aerobic oxidative amination of alkenes

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service