Skip to Content
MilliporeSigma
All Photos(1)

Documents

486248

Sigma-Aldrich

Tris-d11 solution

1 M in D2O, 98 atom % D

Synonym(s):

Tris(hydroxymethyl-d3)amino-d2-methane solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(DOCD2)3CND2
Molecular Weight:
132.20
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

concentration

1 M in D2O

technique(s)

NMR: suitable
bio NMR: suitable

refractive index

n20/D 1.346

density

1.137 g/mL at 25 °C

mass shift

M+11

SMILES string

[2H]OC([2H])([2H])C(N([2H])[2H])(C([2H])([2H])O[2H])C([2H])([2H])O[2H]

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2/i1D2,2D2,3D2,6D,7D,8D/hD2

InChI key

LENZDBCJOHFCAS-RNVFPDRBSA-N

General description

Tris-d11 solution is the isotope-labeled analog of tris(hydroxymethyl)aminomethane. It is suitable for NMR and bio-NMR applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service