Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

164488

Sigma-Aldrich

Sodium deuteroxide solution

30 wt. % in D2O, 99 atom % D

Synonym(s):

Sodium deuteroxide solution, Sodium hydroxide-d

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOD / D2O
CAS Number:
Molecular Weight:
41.00
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

form

liquid

concentration

30 wt. % in D2O

technique(s)

NMR: suitable

refractive index

n20/D 1.4039

mass shift

M+1

SMILES string

[Na+].[O-][2H]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1/i/hD

InChI key

HEMHJVSKTPXQMS-DYCDLGHISA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Zhai et al.
Biochemistry, 53(21), 3486-3501 (2014-05-16)
Two mutations of the phosphodianion gripper loop in chicken muscle triosephosphate isomerase (cTIM) were examined: (1) the loop deletion mutant (LDM) formed by removal of residues 170-173 [Pompliano, D. L., et al. (1990) Biochemistry 29, 3186-3194] and (2) the loop
Oliver Wahl et al.
Journal of pharmaceutical and biomedical analysis, 95, 1-10 (2014-03-19)
For the impurity profiling of the mucolytic and anti-inflammatory drug carbocisteine a high performance liquid chromatographic (HPLC) method using corona charged aerosol detection (CAD) was developed and fully validated following the ICH guideline Q2(R1). The response was linear (R²>0.995) over
Dominique E Williams et al.
Chembiochem : a European journal of chemical biology, 16(10), 1474-1482 (2015-05-09)
With the goal of designing a lysosomal phospholipase mimic, we optimized experimental variables to enhance Ce(IV) -assisted hydrolysis of phosphatidylcholine (PC) liposomes. Our best result was obtained with the chelating agent bis-tris propane (BTP). Similar to the hydrolytic enzyme, Ce(IV)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service