Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.13366

Sigma-Aldrich

Deuterium oxide

deuteration degree min. 99.9% for NMR spectroscopy MagniSolv

Synonym(s):

Deuterium oxide, Heavy water

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
MDL number:
UNSPSC Code:
12142201
EC Index Number:
232-148-9
NACRES:
NA.22
grade:
NMR grade
technique(s):
NMR: suitable
bp:
101 °C/1013 hPa

grade

NMR grade

Quality Level

form

liquid

technique(s)

NMR: suitable

pH

7 (20 °C in H2O)

bp

101 °C/1013 hPa

mp

3.8 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O([2H])[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Application

Deuterium oxide deuteration degree min. 99.9% for NMR spectroscopy MagniSolv. CAS 7789-20-0, pH 7 (H₂O, 20 °C).

Analysis Note

Deuteration degree: ≥ 99.90 %
Performance test (NMR-spectrum): passes test

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service