Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

373842

Sigma-Aldrich

Sodium formate-d

99 atom % D

Synonym(s):

Formic-d acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCO2Na
CAS Number:
Molecular Weight:
69.01
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

form

powder

technique(s)

NMR: suitable
bio NMR: suitable

mp

259-262 °C (lit.)

mass shift

M+1

SMILES string

[Na+].[2H]C([O-])=O

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1/i1D;

InChI key

HLBBKKJFGFRGMU-RWQOXAPSSA-M

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service