Skip to Content
MilliporeSigma
All Photos(1)

Documents

295698

Sigma-Aldrich

Sulfur dioxide

≥99.9%

Synonym(s):

Sulfurous anhydride, Sulfurous oxide, Sulphur dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SO2
CAS Number:
Molecular Weight:
64.06
Beilstein:
3535237
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.26 (21 °C, vs air)

Quality Level

vapor pressure

1779 mmHg ( 21 °C)

Assay

≥99.9%

bp

−10 °C (lit.)

mp

−73 °C (lit.)

density

1.25 g/mL at 25 °C (lit.)

SMILES string

O=S=O

InChI

1S/O2S/c1-3-2

InChI key

RAHZWNYVWXNFOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfur dioxide is primarily used as a feedstock for the production of sulfuric acid (contact process). It is generally used in the synthesis of a variety of sulfur-containing organic compounds like alkyl/aryl sulfonyl chlorides, sulfinates, sultines, and polysulfone.

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Press. Gas Liquefied gas - Skin Corr. 1B

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfur dioxide
Understanding Wine Chemistry, 4(7), 140-148 (2016)
Sulfur dioxide.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
The development and application of sulfur dioxide surrogates in synthetic organic chemistry.
Emmett EJ and Willis MC
Asian Journal of Organic Chemistry, 4(7), 602-611 (2015)
Eliza Harris et al.
Science (New York, N.Y.), 340(6133), 727-730 (2013-05-11)
Global sulfate production plays a key role in aerosol radiative forcing; more than half of this production occurs in clouds. We found that sulfur dioxide oxidation catalyzed by natural transition metal ions is the dominant in-cloud oxidation pathway. The pathway
J Neil Cape et al.
Environment international, 29(2-3), 201-211 (2003-04-05)
The regulation of the emissions of 'traditional' primary air pollutants (fluorides, sulfur dioxide) has changed the pattern of exposure of ecological systems, with greatly reduced exposure close to sources, but with a smaller effect in some remote areas. Measurements show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service