Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z273910

Sigma-Aldrich

SNOOP® gas leak detector

for reaching places with limited accessibility

Synonym(s):

SNOOP® Liquid Leak Detector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.51

technique(s)

gas chromatography (GC): suitable

Looking for similar products? Visit Product Comparison Guide

General description

Nontoxic, nonflammable and leaves no residue. 8 oz, squeezable bottle with tube extendable to 12 in.

Other Notes

For use with volumetric (bubble) flowmeters. Also useful for checking gas delivery system plumbing for leaks. Not recommend for use upstream of capillary GC systems (an electronic leak detector is preferred).

Legal Information

SNOOP is a registered trademark of Swagelok Company

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service