Key Documents
Z146951
Sigma-Aldrich
Lecture-bottle control valve
stainless steel, hose outlet I.D. 1/4 in.
Synonym(s):
Stainless steel lecture-bottle control valve
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
stainless steel
Joint
joint CGA Inlet
joint hose outlet
parameter
-65-250 °F temperature (-54-120 °C)
3,000 psi pressure range (@ 70°F)
hose outlet I.D.
1/4 in.
fitting
CGA 110 male
General description
CGA110, straight-pattern valve for precise flow control of lecture-bottle gas. Requires CGA110 PTFE sealing washer.
Brass control valve shown.
Warning
Gas-control valves do not control pressure in a closed system. Regulator or relief valves should be used to prevent the buildup of unsafe pressures that could lead to explosion.
Choose from one of the most recent versions:
Certificates of Analysis (COA)