Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

295132

Sigma-Aldrich

Chlorine

Chlorine

≥99.5%

Synonym(s):

Chlorine gas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2
CAS Number:
Molecular Weight:
70.91
Beilstein:
3902968
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
bp:
−34 °C (lit.)
vapor pressure:
4800 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.48 (vs air)

Quality Level

vapor pressure

4800 mmHg ( 20 °C)

Assay

≥99.5%

form

gas (compressed)

resistivity

1E9 μΩ-cm, 20°C

pH

1.8 (20 °C, 6.4 g/L)

bp

−34 °C (lit.)

mp

−101 °C (lit.)

SMILES string

ClCl

InChI

1S/Cl2/c1-2

InChI key

KZBUYRJDOAKODT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chlorine gas has been used to generate chlorine atoms via photolysis for molecular halogen photochemical experiments.[1]

It can be used:
  • As a reactant for the extraction of gold from an alluvial material via a pyrometallurgical process.[2]
  • To synthesize carbide-derived carbons (CDC) from zirconium carbide.[3]

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Gas 1 - Press. Gas Compr. Gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zilong Mi et al.
Journal of environmental sciences (China), 37, 200-205 (2015-11-18)
Disinfectants are commonly applied to control the growth of microorganisms in drinking water distribution systems. However, the effect of disinfection on drinking water microbial community remains poorly understood. The present study investigated the impacts of different disinfectants (chlorine and chloramine)
Superactivated carbide-derived carbons with high hydrogen storage capacity.
Sevilla M, et al.
Energy & Environmental Science, 3(2), 223-227 (2010)
Gold extraction by chlorination using a pyrometallurgical process
Ojeda M W, et al.
Minerals Engineering, 22(4), 409-411 (2009)
Halogenation processes of secondary organic aerosol and implications on halogen release mechanisms
Ofner J, et al.
Atmospheric Chemistry and Physics, 12(13), 5787-5806 (2012)
Xinming Li et al.
Nanoscale, 5(5), 1945-1948 (2013-01-30)
We demonstrated the p-type chemical doping by chlorine and nitrate anions to enhance the Schottky junction in the solar cell. Nitrate ions were found to be more effective for reducing the sheet resistance and enlarging the work function of graphene

Articles

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. Akron, Ohio
    • Reviews 2
    • Votes 0
    5 out of 5 stars.

    Easy to use size.

    A strong oxidizer that was needed for the synthesis that came in a small lecture bottle.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service