Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

926779

Sigma-Aldrich

Sulfur dioxide solution

2.0 M in acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SO2
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.06

form

liquid

Quality Level

concentration

2.0 M in acetonitrile

density

0.84113

SMILES string

[S](=O)=O

InChI

1S/O2S/c1-3-2

InChI key

RAHZWNYVWXNFOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfur dioxide solution (2.0 M in MeCN) can be used in electrochemical transformations, including the metal-free synthesis of sulfonamides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Gomi et al.
The Journal of antibiotics, 41(4), 425-432 (1988-04-01)
Structures of new antibiotics SF2446A1, A2, A3, B1, B2 and B3 have been deduced by means of spectral analyses and chemical studies. The structure of SF2446A1 which is a main product of fermentation and has the strongest antimicrobial activity, has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service