Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

241008

Sigma-Aldrich

Phenethylamine

≥99%

Synonym(s):

2-Phenethylamine, β-Phenylethylamine, 2-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
507488
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.18 (vs air)

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

197-200 °C (lit.)

solubility

alcohol: freely soluble(lit.)
diethyl ether: freely soluble(lit.)
water: soluble(lit.)

density

0.962 g/mL at 20 °C (lit.)

functional group

amine
phenyl

SMILES string

NCCc1ccccc1

InChI

1S/C8H11N/c9-7-6-8-4-2-1-3-5-8/h1-5H,6-7,9H2

InChI key

BHHGXPLMPWCGHP-UHFFFAOYSA-N

Gene Information

human ... AOC3(8639)
mouse ... Aoc3(11754)
rat ... Htr2a(29595)

Looking for similar products? Visit Product Comparison Guide

General description

Phenethylamine, a new stimulant designer drug, was analyzed by the liquid chromatography high-resolution mass spectrometry quadrupole-time-of-flight (LC-HRMS-QTOF) method[1].

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Paul et al.
Analytical and bioanalytical chemistry, 406(18), 4425-4441 (2014-05-16)
The development of a liquid chromatography high-resolution mass spectrometry quadrupole-time-of-flight (LC-HRMS-QTOF) method for the analysis of new stimulant designer drugs (e.g. phenethylamine, amphetamine, cathinone and piperazine derivatives) and common drugs of abuse (e.g. ketamine and ritalinic acid) in urine is
Deepak Narang et al.
The Journal of pharmacology and experimental therapeutics, 351(1), 164-171 (2014-08-15)
The trace amine β-phenylethylamine (PEA) is normally present in the body at low nanomolar concentrations but can reach micromolar levels after ingestion of drugs that inhibit monoamine oxidase and primary amine oxidase. In vivo, PEA elicits a robust pressor response
Douwe de Boer et al.
Pharmacy world & science : PWS, 26(2), 110-113 (2004-04-17)
To collect data related to phenethylamine drugs-of-abuse of the 2C-series, to review possible health risks of their use and to discuss legal counter actions of authorities in the European Union (EU). Dutch smartshops. In the period of 1994-2002, all products
Marion Seitter et al.
International journal of food microbiology, 145(2-3), 483-487 (2011-02-22)
In connection with a study on the DNA microarray based detection of genes involved in safety and technologically relevant properties (Seitter (née Resch) et al., 2011), food-associated coagulase-negative staphylococci (CNS) were investigated phenotypically with regard to their ability to bind
Keiko Nakanishi et al.
Forensic science international, 217(1-3), 174-181 (2011-12-14)
Cross-reactivities of 76 kinds of phenethylamine-type designer drugs and related compounds to the urine drug tests Instant-View ™ (IV) (the Methamphetamine (MA) test, the Amphetamine 300 test, and the MDMA test) have been investigated. An on-site urine test kit consisting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service