Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.07047

Sigma-Aldrich

(S)-(-)-1-Phenylethylamine

for the resolution of racemates for synthesis

Synonym(s):

(S)-(-)-1-Phenylethylamine, L-α-Methylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
MDL number:
UNSPSC Code:
12352116
EC Index Number:
220-098-0
NACRES:
NA.22

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

355 °C

potency

980 mg/kg LD50, oral (Rat)
730 mg/kg LD50, skin (Rabbit)

pH

>7 (20 °C in H2O, undiluted)

bp

187 °C/1013 hPa

mp

-10 °C

transition temp

flash point 71 °C

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N[C@@H](C)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m0/s1

InChI key

RQEUFEKYXDPUSK-ZETCQYMHSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.951 - 0.953
Spec. rotation [α²0/D (undiluted): -40.0 - -38.0 °
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service