Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

471283

Sigma-Aldrich

Triethylamine

≥99.5%

Synonym(s):

N,N-Diethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
Beilstein:
605283
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.21
bp:
88.8 °C (lit.)
vapor pressure:
51.75 mmHg ( 20 °C)

vapor density

3.5 (vs air)

Quality Level

vapor pressure

51.75 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

593 °F

expl. lim.

8 %

impurities

≤0.1% (Karl Fischer)

refractive index

n20/D 1.401 (lit.)

pH

12.7 (15 °C, 100 g/L)

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

solubility

water: soluble 112 g/L at 20 °C

density

0.726 g/mL at 25 °C (lit.)

functional group

amine

storage temp.

room temp

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylamine (TEA, Et3N) is an aliphatic amine. Its addition to matrix-assisted laser desorption/ionization (MALDI) matrices affords transparent liquid matrices with enhanced ability for spatial resolution during MALDI mass spectrometric (MS) imaging. A head-space gas chromatography (GC) procedure for the determination of triethylamine in active pharmaceutical ingredients has been reported. The viscosity coefficient of triethylamine vapor over a range of density and temperature has been measured.

Application

Triethylamine has been used during the synthesis of:
  • 5′-dimethoxytrityl-5-(fur-2-yl)-2′-deoxyuridine
  • 3′-(2-cyanoethyl)diisopropylphosphoramidite-5′-dimethoxytrityl-5-(fur-2-yl)-2′-deoxyuridine
  • polyethylenimine600-β-cyclodextrin (PEI600-β-CyD)
It may be used as a homogeneous catalyst for the preparation of glycerol dicarbonate, via transesterification reaction between glycerol and dimethyl carbonate (DMC).

Packaging

18 L packaged in closed-head steel drums; 100 mL, 500 mL, 2 L, 6×500 mL, 4 L packaged in glass bottles; 200 L packaged in Kilo-Lab® steel cylinders

Legal Information

Kilo-Lab is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of glycerol 1,2-carbonate by transesterification of glycerol with dimethyl carbonate using triethylamine as a facile separable homogeneous catalyst.
Ochoa-Gomez JR, et al.
Green Chemistry, 14(12), 3368-3376 (2012)
Nicholas J Greco et al.
Nature protocols, 2(2), 305-316 (2007-04-05)
We describe procedures for the synthesis of a fluorescent pyrimidine analog and its site-specific incorporation into a DNA oligomer. The 5'-protected and 3'-activated nucleoside 4 is synthesized in three steps with an overall yield of 40%. Site-specific incorporation into a
The viscosity of triethylamine vapor at low densities and in the saturated vapor phase.
Teske V, et al.
Fluid Phase Equilibria, 293(2), 190-195 (2010)
GC quantification of cyclopropylamine, diethylamine and triethylamine in active pharmaceutical ingredients.
Raghuram P, et al.
Chromatographia, 71(9-10), 963-966 (2010)
Transparent triethylamine-containing MALDI matrices.
Rubakhin SS and Sweedler JV.
Israel J. Chem., 47(2), 185-193 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service