Skip to Content
MilliporeSigma

8.07334

2-Phenylethylamine

for synthesis

Synonym(s):

2-Phenylethylamine, Phenethylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H11N
CAS Number:
Molecular Weight:
121.18
UNSPSC Code:
12352116
EC Index Number:
200-574-4
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.4 hPa ( 20 °C)

Quality Segment

description

Drug Control: Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

assay

≥99.0% (GC)

form

liquid

autoignition temp.

425 °C

potency

300 mg/kg LD50, oral (Rat)

expl. lim.

1.0-5.5 % (v/v)

drug control

Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

pH

11.5 (20 °C, 4.3 g/L in H2O)

kinematic viscosity

2.40 cSt(23 °C)

bp

200-202 °C/1013 hPa

mp

<-60 °C

transition temp

flash point 81 °C

solubility

63.25 g/L

density

0.96 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NCCc1ccccc1

InChI

1S/C8H11N/c9-7-6-8-4-2-1-3-5-8/h1-5H,6-7,9H2

InChI key

BHHGXPLMPWCGHP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.960 - 0.962
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test


pictograms

CorrosionSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 1

flash_point_f

177.8 °F - closed cup

flash_point_c

81 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library