Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

433810

Sigma-Aldrich

(−)-Bis[(S)-1-phenylethyl]amine

99%

Synonym(s):

(−)-Bis[(S)-α-methylbenzyl]amine, [S-(R*,R*)]-(−)-Bis(α-methylbenzyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5CH(CH3)]2NH
CAS Number:
Molecular Weight:
225.33
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]/D −159°, c = 2 in ethanol

optical purity

ee: ≥99% (GLC)

refractive index

n20/D 1.5525 (lit.)

bp

86 °C/0.05 mmHg (lit.)

density

0.987 g/mL at 25 °C (lit.)

SMILES string

C[C@H](N[C@@H](C)c1ccccc1)c2ccccc2

InChI

1S/C16H19N/c1-13(15-9-5-3-6-10-15)17-14(2)16-11-7-4-8-12-16/h3-14,17H,1-2H3/t13-,14-/m0/s1

InChI key

NXLACVVNHYIYJN-KBPBESRZSA-N

Application

(−)-Bis[(S)-1-phenylethyl]amine can be used as:
  • A key intermediate in the synthesis of phosphoramidite (Feringa) ligands.
  • A reactant in the preparation of C(18)-C(24) unit of a novel macrolide compound lasonolide A.
  • A starting material in the synthesis of tacrine−huperzine A hybrid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis of the C(18)-C(24) unit of lasonolide A
Nowakowski M and Hoffmann HMR
Tetrahedron Letters, 38(6), 1001-1004 (1997)
(R)-2, 2′-Binaphthoyl-(S, S)-di (1-phenylethyl) aminophosphine. scalable protocols for the syntheses of phosphoramidite (feringa) ligands
Organic Syntheses (2008)
Enantioselective synthesis of tacrine-huperzine A hybrids. Preparative chiral MPLC separation of their racemic mixtures and absolute configuration assignments by X-ray diffraction analysis
Camps P, et al.
Tetrahedron Asymmetry, 9(5), 835-849 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service