Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

229679

Sigma-Aldrich

Holmium(III) oxide

powder, 99.999% trace metals basis

Synonym(s):

Rare earth

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ho2O3
CAS Number:
Molecular Weight:
377.86
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: holmium

mp

2330 °C (lit.)

SMILES string

O=[Ho]O[Ho]=O

InChI

1S/2Ho.3O

InChI key

JYTUFVYWTIKZGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Holmium(III) oxide (Ho₂O₃) is a rare earth metal oxide known for its unique optical and electrical properties. It exhibits significant utility as a contrast agent in magnetic resonance imaging (MRI), in wavelength-calibration instruments, and as a catalyst in pyrolysis processes. Additionally, Ho₂O₃ is utilized in producing specialty glasses and ceramics, where it imparts distinctive colors and enhances the material′s properties. Its high melting point and chemical stability make it suitable for demanding environments in both industrial and medical sectors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sibel Ebru Yalcin et al.
Science advances, 6(30), eaaz9708-eaaz9708 (2020-08-25)
Knowledge of the occurrences of water films on minerals is critical for global biogeochemical and atmospheric processes, including element cycling and ice nucleation. The underlying mechanisms controlling water film growth are, however, misunderstood. Using infrared nanospectroscopy, amplitude-modulated atomic force microscopy

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service