Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

203181

Sigma-Aldrich

Dysprosium(III) oxide

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Dy2O3
CAS Number:
Molecular Weight:
373.00
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38161201
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: dysprosium

density

7.81 g/mL at 25 °C (lit.)

SMILES string

O=[Dy]O[Dy]=O

InChI

1S/2Dy.3O

InChI key

NLQFUUYNQFMIJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Peeters et al.
Physics in medicine and biology, 51(6), N127-N137 (2006-03-03)
Susceptibility markers for passive tracking need to be small in order to maintain the shape and mechanical properties of the endovascular device. Nevertheless, they also must have a high magnetic moment to induce an adequate artefact at a variety of
Masoud Salavati-Niasari et al.
Ultrasonics sonochemistry, 17(5), 870-877 (2010-03-23)
Dysprosium carbonates nanoparticles were synthesized by the reaction of dysprosium acetate and NaHCO(3) by a sonochemical method. Dysprosium oxide nanoparticles with average size about 17 nm were prepared from calcination of Dy(2)(CO(3))(3).1.7H(2)O nanoparticles. Dy(OH)(3) nanotubes were synthesized by sonication of
S E Seltzer et al.
Journal of computer assisted tomography, 5(3), 370-374 (1981-06-01)
We used a stepwise approach to identify, design, synthesize, and test new high atomic number particulate contrast agents that would be especially well suited for use with computed tomography (CT). Our goal was to produce extremely radiopaque compounds with highly
Happy et al.
Journal of nanoscience and nanotechnology, 7(3), 907-915 (2007-04-25)
The experimental parameters that control the size and size distribution of dysprosium oxide nanoparticles synthesized by homogeneous precipitation technique have been systematically investigated. The particles were characterized with respect to their size, shape, and thermal decomposition behavior. It was found
B Vengala Rao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(3), 951-953 (2008-04-01)
Emission spectrum of Dy(3+):Ca(4)GdO(BO(3))(3) powder phosphor has been analyzed. Two emission bands at 492 and 588 nm with lambda(exci)=311 nm ((6)H(15/2)-->(4)L(19/2)) have been measured from this phosphor. For the intense yellow emission at 588 nm ((4)F(9/2)-->(6)H(13/2)), its lifetime has been

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service