Skip to Content
MilliporeSigma
All Photos(1)

Documents

590509

Sigma-Aldrich

Terbium(III) oxide

99.99% trace metals basis

Synonym(s):

Diterbium trioxide, Terbium oxide, Terbium sesquioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tb2O3
CAS Number:
Molecular Weight:
365.85
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: terbium

SMILES string

O=[Tb]O[Tb]=O

InChI

1S/3O.2Tb

InChI key

GWYXTVGANSBRNB-UHFFFAOYSA-N

General description

Terbium(III) oxide (Tb2O3) is a rare earth oxide with high thermal stability and ability to absorb and emit light at specific wavelengths. It is widely used as a key component in solid-state lighting, display technologies, and as a dopant in lasers. Additionally, Tb2O3 is utilized in the production of green phosphors for cathode ray tubes (CRTs) and LED lighting, enhancing energy efficiency and color rendering.

Application

  • Terbium (III) Oxide (Tb2O3) Transparent Ceramics by Two-Step Sintering from Precipitated Powder: This research discusses the processing and properties of Terbium(III) oxide in the creation of transparent ceramics, highlighting its potential in various high-tech applications (Hu et al., 2022).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priyam Singh et al.
Nanotechnology, 28(41), 415701-415701 (2017-07-19)
We present novel inorganic-organic hybrid nanoparticles (HNPs) constituting inorganic NPs, NaY

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service