Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
Ho2O3
CAS Number:
Molecular Weight:
377.86
UNSPSC Code:
12352303
EC Index Number:
235-015-3
NACRES:
NA.22
MDL number:
Quality Segment
form
solid
mp
2330 °C
storage temp.
no temp limit
SMILES string
[Ho+3].[Ho+3].[O-2].[O-2].[O-2]
InChI
1S/2Ho.3O/q2*+3;3*-2
InChI key
OWCYYNSBGXMRQN-UHFFFAOYSA-N
Application
- Synthesis of holmium-oxide nanoparticles for near-infrared imaging and dye-photodegradation: Discusses the synthesis and application of holmium oxide nanoparticles for use in near-infrared imaging and as agents for photodegradation of dyes. (Zhu et al., 2022).
Analysis Note
Assay (calculated on calcined substance): ≥ 99.5 %
Loss on ignition (800 °C): ≤ 2.0 %
Identity (ICP-OES): passes test
Loss on ignition (800 °C): ≤ 2.0 %
Identity (ICP-OES): passes test
Still not finding the right product?
Explore all of our products under Holmium(III) oxide
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.