Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

176796

Sigma-Aldrich

Sulfuric acid-d2 solution

96-98 wt. % in D2O, 99.5 atom % D

Synonym(s):

Dideuterosulfuric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2SO4/D2O
CAS Number:
Molecular Weight:
100.09
EC Number:
MDL number:
UNSPSC Code:
12352322
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99.5 atom % D

Assay

≥95.5—102.5% (Titration by NaOH)

form

liquid

concentration

96-98 wt. % in D2O

technique(s)

NMR: suitable

refractive index

n20/D 1.4175

mass shift

M+2

storage temp.

room temp

SMILES string

[2H]OS(=O)(=O)O[2H]

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)/i/hD2

InChI key

QAOWNCQODCNURD-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

D2SO4, isotopically labeled acid solution.

Application

Sulfuric acid-d2 solution can be used as an internal standard for 1H-NMR spectroscopy analysis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor Ying Zhang et al.
Physical chemistry chemical physics : PCCP, 21(35), 19147-19152 (2019-08-23)
Understanding how electrolyte composition controls electrocatalytic reactions requires molecular-level insight into electrode/electrolyte interaction. Perhaps the most basic aspect of this interaction, the speciation of the interfacial ion, is often controversial for even relatively simple systems. For example, for Pt(111) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service