Documentos clave
Z201022
Sigma-Aldrich
Spilfyter® chemical classifier
pkg of 1 ea
Seleccione un Tamaño
1 EA
US$ 229,00
US$ 229,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 229,00
About This Item
Código UNSPSC:
47131905
NACRES:
NB.35
US$ 229,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
envase
pkg of 1 ea
Descripción general
Quickly distinguish unknown spilled or stored chemicals. Classifier consists of a test strip with various positioned color indicators that are sensitive to different chemical classes (e.g., pH, oxidizers, fluorides, organics, and chlorides). Strips are sealed against premature exposure by a protective film.
Each kit includes six classifiers, 120 ft PP string, and a laminated color-identification chart. Shipped in an airtight tube.
Información legal
Spilfyter is a registered trademark of National Packaging Services Corp.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)