Skip to Content
Merck
All Photos(1)

Key Documents

Z370541

Sigma-Aldrich

Nalgene® bottle-top sterile filter units

filter capacity 150 mL, pore size 0.2 μm, fits 33 mm bottle neck

Synonym(s):

bottletop filtration, rapid-flow filter, sterile culture media filter, vacuum filtration

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
$144.00

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
12 EA
$144.00

About This Item

UNSPSC Code:
41104914
NACRES:
NB.24

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polyethersulfone membrane
polystyrene housing

sterility

sterile

feature

cap
graduations

packaging

case of 12
pack of 12

manufacturer/tradename

Nalgene® 596-3320

diam.

50 mm

filter capacity

150 mL

funnel capacity

150 mL

neck diam.

33 mm

pore size

0.2 μm

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Nalgene® bottle-top sterile filter units are suitable for the filtration of cell culture media, additives, serum, and buffers.

Features and Benefits

Nalgene® bottle-top sterile filter units have the following benefits:
  • Polyethersulfone (PES) membrane is best suited for tissue culture.
  • Faster results and less waiting time.
  • Non-pyrogenic and endotoxin free.
  • PES exhibits low protein binding, resulting in low chances of loss of protein from media.
  • PES is hydrophilic and thus does not require external wetting agents or surfactants.
  • PES offers higher aqueous flow rates.
  • PES has greater strength, capacity, and chemical resistance than cellulose acetate, nitrocellulose, or nylon.
  • Polystyrene body with graduations.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service