Skip to Content
Merck
All Photos(1)

Documents

TMO597-4520

Nalgene® bottle-top sterile filter units

filter capacity 1000 mL, polysulfone, pore size 0.2 μm, case of 12 ea

Synonym(s):

bottletop filtration, rapid-flow filter, sterile culture media filter, vacuum filtration

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

material

polyethersulfone membrane
polystyrene housing
polysulfone

sterility

sterile

feature

cap
graduations

packaging

case of 12 ea

manufacturer/tradename

Nalgene 597-4520

pore size

0.2 μm pore size

Looking for similar products? Visit Product Comparison Guide

Application

Nalgene® bottle-top sterile filter units are suitable for the filtration of cell culture media, additives, serum, and buffers.

Features and Benefits

Nalgene® bottle-top sterile filter units have the following benefits:
  • Polyethersulfone (PES) membrane is best suited for tissue culture.
  • Faster results and less waiting time.
  • Non-pyrogenic and endotoxin free.
  • PES exhibits low protein binding, resulting in low chances of loss of protein from media.
  • PES is hydrophilic and thus does not require external wetting agents or surfactants.
  • PES offers higher aqueous flow rates.
  • PES has greater strength, capacity, and chemical resistance than cellulose acetate, nitrocellulose, or nylon.
  • Polystyrene body with graduations.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service