SAE0157
Potassium Acetate Buffer Solution
100 mM, pH 5.0, 0.2 μM filtered
Select a Size
$58.50
Select a Size
About This Item
$58.50
Recommended Products
General description
Application
- Buffering urine in drug hydrolysis procedures when using limpet β-glucuronidase enzymes such as SRE0093, SRE0095, G8132 & G2174
- The enzymatic assay of β-glucuronidase from Helix pomatia and Bovine Liver (Protocol)
- Use as a general laboratory pH 5.0 acetate buffering reagent.
Features and Benefits
- 0.2 μm filtered
- pH 5.0 for optimal limpet β-glucuronidase enzyme drug hydrolysis
The potassium acetate buffer solution was developed specifically for use with the recombinant limpet β-glucuronidase enzyme products SRE0093 and SRE0095. The buffer is free of phosphate, detergents, carbohydrates and other components that may interfere with analysis methods.
Storage and Stability
Other Notes
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service