Key Documents
HS2134A
Floating Microtube Rack
to hold, 20 x 1,5/2.0 mL tubes, round white polypropylene
Select a Size
4 EACH
$63.60
$63.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
4 EACH
$63.60
About This Item
UNSPSC Code:
41122800
NACRES:
NB.12
$63.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
round white polypropylene
feature
to hold 20 x 1,5/2.0 mL tubes
packaging
pack of 4 ea
manufacturer/tradename
Heathrow Scientific HS2134A
Related Categories
General description
The Floating Microtube Rack can be used on the bench, with water baths or ice containers. The Rack keeps microtubes afloat in a water bath or can be pushed into crushed ice. It is perfect for controlling sample temperature.
Choose from one of the most recent versions:
Certificates of Analysis (COA)