Key Documents
HS2132A
Floating Microtube Rack
to hold, 16 x 1,5/2.0 mL tubes, square white polypropylene
Select a Size
4 EACH
$63.60
$63.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
4 EACH
$63.60
About This Item
UNSPSC Code:
41122800
NACRES:
NB.12
$63.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
square white polypropylene
feature
to hold 16 x 1,5/2.0 mL tubes
packaging
pack of 4 ea
manufacturer/tradename
Heathrow Scientific HS2132A
Related Categories
General description
The Floating Microtube Rack can be used on the bench, with water baths or ice containers. The Rack keeps microtubes afloat in a water bath or can be pushed into crushed ice. It is perfect for controlling sample temperature.
Features and Benefits
  • Square Shape Rack
  • 1.5/2.0 mL Tubes, 16 Wells (10.9 mm well dia.)
  • Use on the bench, with water baths or ice containers
  • Keeps microtubes afloat in a water bath or can be pushed into crushed ice — perfect for controlling sample temperature
  • Attach legs so rack stands upright on bench top
  • Detach legs so samples can be quickly removed by pushing down onto any surface
  • Not recommended for autoclaving
Choose from one of the most recent versions:
Certificates of Analysis (COA)