Key Documents
BAF187420001
PrepSafe Floating Microcentrifuge Tube Rack
Holds 28 x 1.5-2 mL tubes, polypropylene
Select a Size
1 EA
$31.40
$31.40
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$31.40
About This Item
UNSPSC Code:
41122804
NACRES:
NB.22
$31.40
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
feature
Holds 28 x 1.5-2 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18742-0001
size
178x64x mm
General description
Protect your samples. Hold 1.5ml and 2.0ml microcentrifuge tubes securely in a water bath or on the benchtop, even if tipped over. Immobilizing tabs allow tubes to be stored securely and ensure that the caps stay locked in place. This floating rack has 28 numbered locations so samples can easily be identified. Raised handle allows rack to be lifted without agitating samples. Patent pending.
Legal Information
PrepSafe is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)