Key Documents
CLS4395
Corning® Costar® HTS Transwell® cell culture plates
tissue-culture treated, sterile, reservoir, pack of 48 ea
Select a Size
48 EA
$381.00
$381.00
Estimated to ship onMay 28, 2025
Select a Size
Change View
48 EA
$381.00
About This Item
UNSPSC Code:
41122107
NACRES:
NB.13
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
$381.00
Estimated to ship onMay 28, 2025
sterility
sterile
packaging
pack of 48 ea
manufacturer/tradename
Corning 4395
binding type
Tissue Culture (TC)-treated surface
General description
This allows cell culture on the membranes of all 96 well HTS Transwell plate types within a relatively large amount of medium. The tray provides optimum culture conditions across the whole plate and good buffering against pH changes and nutrient depletion. Trays contain a removable stabilizer which reduces medium sloshing during manipulation. Reservoir trays are used during the active cell growth and feeding phase prior to transferring the HTS Transwell system to a receiver plate which segregates the individual inserts for the final assay step. This reservoir tray is tissue culture treated and sterile for applications requiring co-cultures with the same or different cell types.
Legal Information
Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)